Learn More
Kingfisher Biotech Inc Bovine GM-CSF ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY1509B004
Encompass_Preferred
The Bovine GM-CSF ELISA contains capture antibody standard and detection antibody for development of a Bovine GM-CSF ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CSF2 GMCSF colony stimulating factor 2 CSF. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5506