Learn More
Biomatik Corporation Bovine Growth Hormone, Gh Elisa Kit
Supplier:  Biomatik Corporation EKC31643
Encompass_Preferred
Bovine Growth Hormone, Gh Elisa Kit for detecting Growth Hormone 1 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.25-50ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01246; GH1. Target Synonyms: GH1; GH; Somatotropin; Growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4366
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.