Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bovine herpesvirus-1 Glycoprotein B Ab ELISA Kit

Catalog No. 89032337 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-337 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-337 Supplier Abnova™ Supplier No. KA4889
Add to Cart
Add to Cart

Enzyme-linked immunosorbent assay for the detection of antibodies to Bovine herpesvirus-1 (BHV-1) glycoprotein B.

Intended for detection of specific antibodies in the sample using blocking ELISA technique (e.g. solid phase coated with specific antigen - antibody from the sample - labeled antibody against the specific antigen on the solid phase).

Specifications

Description Bovine herpesvirus-1 Glycoprotein B Ab ELISA Kit is an enzyme-linked immunosorbent assay for the detection of antibodies to Bovine herpesvirus-1 (BHV-1) glycoprotein B.
Conjugate HRP
Detection Method Colorimetric
Product Type Bovine herpesvirus-1 Glycoprotein B Ab ELISA Kit
Sample Type Milk, Plasma, Serum
Target Species Bovine
For Use With (Application) Qualitative
Kit Contents Microtitre Plate: 96 wells, Negative Control Serum: 2mL, Positive Control Serum - 2mL, Conjugate: 30mL, Sample Diluent 12 (20mL), TMB-Complete 3 (28mL), Wash Solution: 20x (75mL), Stop Solution: Acid solution ready to use (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Storage Requirements Store the kit at 4°C. Do not freeze.
Target Bovine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.