Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine IFN-gamma Recombinant Protein

Catalog No. ENRBOIFNGI
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRBOIFNGI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRBOIFNGI Supplier Invitrogen™ Supplier No. RBOIFNGI
Add to Cart
Add to Cart

Recombinant Protein

Mature bovine IFNγ was expressed in Pichia pastoris, X-33. GenBank® Accession # AAB64477, amino acids 24 - 166. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: QGQFFREIEN. >95% by SDS-PAGE analysis. Activity: The biological activity of recombinant bovine IFNγ was determined in a viral challenge assay using VS virus on MDBK cells. Plates were stained with crystal violet, a visual CPE determination was performed and the dye was then solubilized and absorbance was read. Uses: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution: Reconstitute lyophilized material with 0.5 mL of distilled or deionized water. Dilute in a buffer containing carrier protein, aliquot, and store at = -70°C. Avoid multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing = 10% FCS are suitable buffers.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07353
Concentration 0.825mg/mL
For Use With (Application) ELISA Standard, Bioactivity, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 281237
Molecular Weight (g/mol) 16.9 kDa
Name Bovine IFN-gamma
Purification Method Purified
Quantity 25 μg
Source Yeast
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Common Name IFN gamma
Gene Symbol IFNG
Biological Activity The biological activity of recombinant bovine IFN-gamma was determined in a viral challenge assay using VSV on MDBK cells. Plates were stained with crystal violet, a visual CPE determination was performed and the dye was then solubilized and absorbance was read. Treatment of MDBK cells with 6 ng/mL of bovine IFN-gamma causes 25% Cytoprotection in the assay.
Product Type Protein
Conjugate Unconjugated
Species Cattle/Bovine
Recombinant Recombinant
Sequence Bovine IFN?, amino acids 24 - 166 (Accession # AAB64477).
Content And Storage -80° C, Avoid Freeze/Thaw Cycles
Activity The biological activity of recombinant bovine IFN-gamma was determined in a viral challenge assay using VSV on MDBK cells. Plates were stained with crystal violet, a visual CPE determination was performed and the dye was then solubilized and absorbance was read. Treatment of MDBK cells with 6 ng/mL of bovine IFN-gamma causes 25% Cytoprotection in the assay.
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE analysis
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.