Learn More
Cygnus Technologies Bovine IgG ELISA Kit, 96 Well Plate Format
Supplier:  Cygnus Technologies F070
Encompass
This kit is intended to quantitate bovine IgG contamination in various types of samples throughout the purification process. The colorimetric endpoint can be read within three hours on a conventional microtiter plate reader to a sensitivity of <250pg/mL of bovine IgG.
Catalog No. NC0199188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.