Learn More
Kingfisher Biotech Inc Bovine IL-1 Receptor Antagonist Biotinylated Recombinant Protein
Supplier: Kingfisher Biotech Inc RPB1885B010
The Bovine IL-1 Receptor Antagonist (IL-1ra) Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-1 Receptor Antagonist (IL-1ra) Biotinylated applications are for cell culture. Bovine IL-1 Receptor Antagonist (IL-1ra) Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-1 Receptor Antagonist (IL-1ra) Biotinylated Specifications: (Molecular Weight: 17.2 kDa) (Amino Acid Sequence: HPLGKRRCEM QAFRIWDVNQ KIFYLRNNQL VAGYLQGPNT KLEEKIDVVP IEPHTMFLGI HGGKLCLACV KSGDEIKLKL EAVNITDLNQ NREQDKRFAF IRFDNGPTTS FESAACPGWF LCTSLEADQP VGLTNMPTEA LKVTKFYFQQ D (151)) (Gene ID: 281860). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.