Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine IL-6 Recombinant Protein

Catalog No. PIRBOIL6I
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRBOIL6I 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRBOIL6I Supplier Invitrogen™ Supplier No. RBOIL6I
Add to Cart
Edge
Add to Cart

Recombinant Protein

For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Reconstitute with 0.5 mL of distilled water.

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26892
For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 280826
Molecular Weight (g/mol) 22 kDa
Name Bovine IL-6
Quantity 20 μg
Gene Alias B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6
Common Name IL-6
Gene Symbol IL6
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Content And Storage 4°C
Expression System P. pastoris
Form Lyophilized
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.