Learn More
Kingfisher Biotech Inc Bovine IL-8 Biotinylated Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RPB1802B010
Encompass
The Bovine IL-8 Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-8 Biotinylated applications are for cell culture. Bovine IL-8 Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-8 Biotinylated Specifications: (Molecular Weight: 9.1 kDa) (Amino Acid Sequence: AVLSRMSTEL RCQCIKTHST PFHPKFIKEL RVIESGPHCE NSEIIVKLTN GNEVCLNPKE KWVQKVVQVF VKRAEKQDP) (Gene ID: 280828). For research use only. Made in the USA
Catalog No. 50-253-2368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.