Learn More
Bioworld Bovine Immunoglobulin G Liquid >= 97%, 10mg/ml, 1000 g
Supplier:  Bioworld 220700274
Encompass_Preferred
The immunoglobulin preparations are supplied as a liquid. Purity levels are >97% IgG as analyzed by SDS-Page. These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps, or as a protein stabilizer blocker in a number of immunoassays. These preparations are also useful as IgG control solutions for biochemical analytical tests. Immunoglobulin preparations are provided at pH 7.2 ± 0.2. Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-253-950
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.