Learn More
American Research Products Inc BoVine Interleukin 2 IL 2 ELI
Supplier:  American Research Products Inc CSBE09810B
Encompass
BoVine Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30188