Learn More
Kingfisher Biotech Inc Bovine Osteopontin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2089B005
Encompass
The Bovine Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Bovine Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Bovine Osteopontin Specifications: (Molecular Weight: 29.2 kDa) (Amino Acid Sequence: LPVKPTSSGS SEEKQLNNKY PDAVATWLKP DPSQKQTFLA PQNSVSSEET DDNKQNTLPS KSNESPEQTD DLDDDDDNSQ DVNSNDSDDA ETTDDPDHSD ESHHSDESDE VDFPTDIPTI AVFTPFIPTE SANDGRGDSV AYGLKSRSKK FRRSNVQSPD ATEEDFTSHI ESEEMHDAPK KTSQLTDHSK ETNSSELSKE LTPKAKDKNK HSNLIESQEN SKLSQEFHSL EDKLDLDHKS EEDKHLKIRI SHELDSASSE VN (262)) (Gene ID: 281499). For research use only. Made in the USA
Catalog No. 50-253-2458
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.