Learn More
Biomatik Corporation Bovine Pigment Epithelium Derived Factor (PEDF) ELISA Kit
Supplier:  Biomatik Corporation EKU06637
Encompass_Preferred
Bovine Pigment Epithelium Derived Factor (PEDF) ELISA Kit for detecting PEDF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q95121. Gene ID: 281386. Target Synonyms: SERPINF1; EPC1; PIG35; SDF3; Serpin F1; Cell proliferation-inducing gene 35; Serpin Peptidase Inhibitor,Clade F Member 1 (Alpha-2 Antiplasmin) ; Stromal Cell Derived Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6119
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.