Learn More
Description
- Bovine Pituitary Extract (BPE) is made from fresh, flash-frozen pituitary glands from young, healthy cattle between the ages of 18 to 22 months
- The cattle are born, raised and slaughtered in New Zealand from a USDA approved source
- New Zealand cattle are internationally recognized as free from bovine spongiform encephalopathy (BSE, mad-cow disease) and have no history of blue tongue disease
- BPE continues to be the most successful of the tissue-sourced extracts to date and allows researchers to study normal primary cell types including stem cells in proliferating cultures
- Formulated at 14 mg/mL crude protein, this aqueous BPE has a full spectrum of mitogens and differentiation factors
- The Bovine Pituitary Extract is not lyophilized, as this process has been shown to reduce biological activity
- For cell culture, the optimal medium supplementation rate is 35 μg/mL BPE based on growth tests on human mammary epithelial cells, epidermal keratinocytes, prostatic and bronchial epithelial cells
- This product has been sterile-filtered prior to shipping
Specifications
Specifications
| Content And Storage | Store at -20°C, do not thaw and refreeze |
| Concentration | 14 mg/mL |
| Form | Liquid |
| Sterility | Sterile |
| For Use With (Application) | Cell Culture |
| Quantity | 10 mL |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.