Learn More
Biomatik Corporation Bovine Relaxin (RLN) ELISA Kit
Supplier:  Biomatik Corporation EKU07059
Encompass_Preferred
Bovine Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 24.69-2000pg/ml. Sensitivity: 9.25pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.