Learn More
Kingfisher Biotech Inc Bovine CTLA-4 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1411B100
Encompass_Preferred
The Bovine sCTLA-4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine sCTLA-4 applications are for cell culture, ELISA standard, and Western Blot Control. Bovine sCTLA-4 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine sCTLA-4 Specifications: (Molecular Weight: 13.8 kDa) (Amino Acid Sequence: FSKGMNVTQP PVVLASSRGV ASFSCEYESS GKADEVRVTV LREAGSQVTE VCAGTYMVED ELTFLDDSTC IGTSRGNKVN LTIQGLRAMD TGLYVCKVEL MYPPPYYVGI GNGTQIYVID PEPCPDSD (128)) (Gene ID: 281732). For research use only. Made in the USA
Catalog No. 50-253-3276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.