Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bovine Serum Albumin
GSA_VA

Catalog No. 5217100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-171-00G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 52-171-00G Supplier Tocris Bioscience™ Supplier No. 5217/100G
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Blocking buffer for IHC, ICC, ELISA and Western blotting; used to block non-specific antibody binding

Bovine Serum Albumin is a BSA (bovine serum albumin) is a widely used blocking reagent for use in immunohistochemistry (IHC), Immunocytochemistry (ICC), ELISAs and Western blotting. Used to block non-specific binding of antibodies. Can also be used as a protein concentration standard in Bradford assay for protein quantification. BSA is also used in cell culture.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Purity 98%
Content And Storage Store at +4°C
Product Type Bovine Serum Albumin
Quantity 100 g

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.