Learn More
Proliant BOVINE SERUM ALBUMIN 250G
Supplier:  Proliant 68100250G
Encompass_Preferred
Proliant Biologicals’ Albumin, Bovine, pH 7.0, Standard Grade is a high purity, lyophilized “Fraction V” powder derived from the plasma of USDA-inspected healthy animals from abattoirs located in the USA. It is an economical choice that is deal for most diagnostic and research applications. These applications include, but are not limited to, Immunoassays (EIA, RIA, etc.), Serology, and Mammalian Cell Culture.
Catalog No. 50-109-0662
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.