Learn More
Proliant BOVINE SERUM ALBUMIN 500G
Supplier:  Proliant 68700500G
Encompass_Preferred
Proliant Biologicals’ Albumin, Bovine, Reagent Grade (Fatty-Acid Free) is a high purity, lyophilized “Fraction V” powder derived from the plasma of USDA-inspected healthy animals from abattoirs located in the USA. It is ideal for the most sensitive diagnostic and research applications. These applications include Nucleic Acid-Based Assays, Immunoassays (RIA, EIA, etc.), Immunohematology and Serology.
Catalog No. 50-109-0677
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.