Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bovine Serum Albumin Native Protein
SDP

Catalog No. NBP196904 Shop All R&D Systems Products
Change view
Click to view available options
1mg; Texas Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP196904 1mg; Texas Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196904 Supplier Novus Biologicals™ Supplier No. NBP196904
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Bovine Serum Albumin. Source: Bovine Serum The Bovine Serum Albumin Native Protein is derived from Bovine Serum. The Bovine Serum Albumin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.

Specifications

For Use With (Application) Dot Blot
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000)
Gene ID (Entrez) 280717
Name Bovine Serum Albumin protein
Storage Requirements 4°C in the dark.
Cross Reactivity Bovine
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.