Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Bovine Serum Albumin Native Protein
SDP

Catalog No. NB815302 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB815302 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB815302 Supplier R&D Systems™ Supplier No. NBP19705810MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Native Protein

Specifications

Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene ID (Entrez) 280717
Quantity 10 mg
Storage Requirements Store at 4°C.
Gene Alias albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbol ALB
Reconstitution Reconstitute with 1.0 ml deionized water.
Inhibitor Properties This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.
Research Category Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Product Line Novus
Purity or Quality Grade Multi-step
Protein Bovine Serum Albumin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.