Learn More
Kingfisher Biotech Inc Bovine ICAM-1 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1127B005
Encompass
The Bovine sICAM-1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine sICAM-1 applications are for cell culture, ELISA standard, and Western Blot Control. Bovine sICAM-1 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine sICAM-1 Specifications: (Molecular Weight: 50.0 kDa) (Amino Acid Sequence: GISIHPSKAI IPRGDSLTVN CSNSCDQKST FGLETVLIKE EVGRGDNWKV FQLRDVQEDI ELFCYSNCHK EQTIASMNLT VYWFPEHVEL APLPLWQPVG EELNLSCLVS GGAPRAHLSV VLLRGEEELG RQPVGKGEPA KVMFTVQSRR EDHGTNFSCR WELDLRSQGL ELFQNTSAPR KLQTYVLPSI DPHLEVPPIV EVGSRWPVNC TLDGLFPASD AKVYLVLGDQ KLESNITYDG DSVLAKAWME ENEEGTHSLK CSVTLGEVSR RTQENVTVYS FPLPTLTLSP PEVSEWTTVT VECVTRDGAV VKLNGTSAVP PGPRAQLKLN ASASDHRSNF SCSAALEIAG QVVHKHQTLE LHVLYGPRLD QRDCPGNWTW QEGSEQTLKC EAQGNPIPKL NCSRKGDGAS LPIGDLRPVR REVAGTYLCR ATSARGRVTR EVVLNVLHGQ NIL (453)) (Gene ID: 281839). For research use only. Made in the USA
Catalog No. 50-253-3642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.