Learn More
Kingfisher Biotech Inc Bovine Thrombopoietin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1134B005
Encompass
The Bovine Thrombopoietin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine Thrombopoietin applications are for cell culture, ELISA standard, and Western Blot Control. Caprine VEGF-A yeast-derived recombinant protein can be purchased in multiple sizes. Bovine Thrombopoietin Specifications: (Molecular Weight: 34.9 kDa) (Amino Acid Sequence: SPAPPACDPR LLNKLLRDSH VLHSRLSQCP DVNPLSTPVL LPVVDFSLGE WKTQTEQTKA QDVLGTTTLL LEAVMTARGQ LGPTCLSSLL VQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSAIFLS FQQLLRGKVR FLLLVVGPSL CAKRAPPATA VPGSISPLLT LNKLPNRTSG LLETNSSVSA RTTGFGLPNR LQGFRAKIPG LLNQTSRSLG QIPGHLNGTQ GPLSGVHGLF PGPSPRALGA PDIPLGTSDM GSLPPYLQPG DSPSPAHPPP GQYTLFSPSP TSPIPTVHLQ PLLPDPSAIT PNSSSSLLVA AHSHFRNLSQ EE (332)) (Gene ID: 538672). For research use only. Made in the USA
Catalog No. 50-253-3622
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.