Learn More
American Research Products Inc BoVine Thyroid Stimulating Hor
Supplier: American Research Products Inc CSBE17548B
BoVine Thyroid Stimulating Hormone (TSH) ELISA Kit from CUSABIO Detection Range 0 9 uIU/mL-60 uIU/mL Sensitivity 0 225 IU/mL Sample serum plasma tissue homogenates Method Sandwich