Learn More
Ampco Safety Tools Box End Wrench (Metric)
SDP
Catalog No. 19173428 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Drive Size:
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
32 mm
33 mm
34 mm
35 mm
36 mm
37 mm
38 mm
41 mm
42 mm
46 mm
47 mm
50 mm
52 mm
54 mm
55 mm
56 mm
58 mm
60 mm
63 mm
65 mm
67 mm
70 mm
71 mm
75 mm
77 mm
80 mm
82 mm
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Drive Size
19-173-428 42 mm
19-173-403 14 mm
19-173-404 15 mm
19-173-405 16 mm
19-173-406 17 mm
19-173-407 18 mm
19-173-408 19 mm
19-173-409 20 mm
19-173-410 21 mm
19-173-411 22 mm
19-173-412 23 mm
19-173-413 24 mm
19-173-414 25 mm
19-173-415 26 mm
19-173-416 27 mm
19-173-417 28 mm
19-173-418 29 mm
19-173-419 30 mm
19-173-420 32 mm
19-173-421 33 mm
19-173-422 34 mm
19-173-423 35 mm
19-173-424 36 mm
19-173-425 37 mm
19-173-426 38 mm
19-173-427 41 mm
19-173-429 46 mm
19-173-430 47 mm
19-173-431 50 mm
19-173-432 52 mm
19-173-433 54 mm
19-173-434 55 mm
19-173-435 56 mm
19-173-436 58 mm
19-173-437 60 mm
19-173-438 63 mm
19-173-439 65 mm
19-173-440 67 mm
19-173-441 70 mm
19-173-442 71 mm
19-173-443 75 mm
19-173-444 77 mm
19-173-445 80 mm
19-173-446 82 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-428 Supplier Ampco Safety Tools Supplier No. 4450
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Box End Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty
Limited Lifetime Warranty
Specifications
Product Type Box End Wrench
Drive Size 42 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.