Learn More
Strem, An Ascensus Company CAS 1416819-91-4. 100mg. 2-[(4S)-4-tert-Butyl-45-dihydro-2-oxazolyl]-5-(trifluoromethyl)pyridine 98% (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 077124100MG
Encompass_Preferred
CAS 1416819-91-4. 100mg. 2-[(4S)-4-tert-Butyl-45-dihydro-2-oxazolyl]-5-(trifluoromethyl)pyridine 98% (99% ee). . Molecular Weight 272.30. Molecular Formula C13H15F3N2O. Color/form white to yellow pwdr. Strem 07-7124. http//www.strem.com/catalog/v/07-7124/
Catalog No. 50-211-0701
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.