Learn More
Tecan Protein Fragment BoxB from HMGB1, human & mouse, LPS-free
Supplier:  Tecan REHM052
Encompass_Preferred
BoxB from HMGB1, human & mouse, with tag, LPS-free - 100 ug . BoxB is one of the DNA binding domains of HMGB1 protein (HMG boxes). Structure-function analysis revealed that the cytokine activity of HMGB1 can be reproduced by recombinant BoxB. It recapitulates the cytokine activity of full length HMGB1 and efficiently activates macrophages to release tumor necrosis factor (TNF) and other proinflammatory cytokines. BoxB from HMGB1 is produced in E.coli from an expression plasmid coding for the mammalian sequence, which is totally identical in human and mouse, preceded by four amino acids (MARI) that have been added for the purpose of expression in bacteria, both of which have counterparts in other eukaryotic regulatory proteins. Distributed by IBL. Research Use Only.
Catalog No. 50-130-6191
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.