Learn More
Eisco™ Boyle Law Apparatus
SCI_ED
Catalog No. S24873 Shop All Eisco Products
Change view
Click to view available options
Boyle Law Apparatus
Catalog No.
S24873 Boyle Law Apparatus
1 options
Catalog No. S24873 Supplier Eisco™ Supplier No. PH0142GL
Add to Cart
Add to Cart
For demonstration of relationship between pressure and volume of gas.
  • With 29mm dia. graduted cylinder with piston and screw valve
  • Cylinder acts upon manometer through narrow passage
  • 60mm dia. manometer scale range -1000 to +3000 hPa with 50hPa interval
  • Gas temperature in cylinder read through digital thermometer in°C
Specifications
Product Type Boyle's Law Apparatus
Sensor Types Manometer, Thermometer
For Use With (Application) Demonstrate the relationship between pressure and volume of gas
Required Accessories 1.5 V AA Battery