Learn More
Boyle's Law and Absolute Zero
SDPSCI_ED
Catalog No. S96823 Shop All Science First Products
Change view
Click to view available options
Boyles Law and Absolute Zero
Catalog No.
S96823 Boyles Law and Absolute Zero
1 options
Catalog No. S96823 Supplier Science First Supplier No. 6112308
Includes
Instructions.
Visualize the basic relationshiop between pressure, volume, and temperature of gases.
  • To show Boyle's Law connect the calibrated syringe to the pressure gauge. View that the corresponding pressure change as the volume of gas in the syringe is varied. Graph results.
  • To show Absolute Zero connect a metal sphere to the pressure gauge and immerse the sphere in water of different temperatures. Read the pressure changes directly from the scale, graph temperature versus pressure, and extrapolate to determine.
Specifications
For Use With (Application) Visualize the basic relationship between pressure, volume and temperature of Gases
Includes Instructions
Product Type Boyle's Law and Absolute Zero Demonstration Aid