Learn More
Eisco™ Boyle's Law Apparatus
SCI_ED
Catalog No. S02912 Shop All Eisco Products
Change view
Click to view available options
Product Type: Boyle's Law Demonstration Apparatus
Catalog No.
S02912 Product Type: Boyle's Law Demonstration Apparatus
1 options
Catalog No. S02912 Supplier Eisco™ Supplier No. PH0147
Add to Cart
Add to Cart
The Eisco Labs Boyle's Law Apparatus includes a horizontally mounted marked acrylic tube, gas tap, pressure gauge, and a rubber stopper on screw threads. It provides a hands-on experience to understand Boyle's Law, showcasing the relationship between pressure and volume in gases.
  • Includes a horizontally mounted marked acrylic tube, gas tap, pressure gauge, and a rubber stopper on screw threads
  • Tube length can be adjusted using the rubber stopper
  • Pressure can be read on the gauge provided. Gas tap attached so gas can be added easily
  • No oil required
  • Perfect for any classroom and useful for a variety of experiments
Specifications
For Use With (Application) Boyles Law Apparatus
Includes Acrylic tube, gas tap, pressure gauge, and a rubber stopper on screw threads
Product Type Boyle's Law Demonstration Apparatus