Learn More
OTT Hydromet Adcon - BP1
Supplier:  OTT Hydromet 200.733.111
Encompass_Preferred
High accurate Barometric Pressure Sensor. Temperature dependencies and non-linearities of the sensor are mathematically compensated for, up to 80°C. The sensor has a very low power consumption and produces an analogue output signal of 0,1 ... 2,5VDC. The piezoresitive sensor element is fully temperature compensated and performs with accuracy better than 0,1%FS across the whole range. According to WMO requirements the sensor itself is encapsulated in a robust polycarbonate housing, which will protect the sensor element from erroneous readings caused by wind gusts. A high-capacity Goretex vent makes sure that pressure inside the case equals outside pressure, and that water ingress is prevented. The sensor box is mounted to the pole of a weather station by means of a mounting bracket and clamp supplied with the product. A shielded 2m cable with a 7-pin Binder jack connects the probe to any RTU of the A723, A733 and A75x product families.
Catalog No. NC1919666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.