Learn More
MilliporeSigma™ Supelco™ BPE-DNPH SPE Cartridge, Bed Wt. 130 mg 2-BPE/270 mg DNPH-coated silica
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Basis Weight | 130 mg 2-BPE/270 mg DNPH-coated silica |
| Material | 2-BPE/DNPH-coated silica |
| For Use With (Application) | Industrial Hygiene |
| Quantity | 50/Pk. |
| Type | SPE Column |
| Volume (Metric) | 3 mL |