Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561743 Supplier Invitrogen™ Supplier No. PA561743
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEEVDQELIE DSQWEEILKQ PCPSQYSAIK EEDLVVWVDP LDGTKEYTEG LLDNVTVLIG IAYEGKAIAG VINQPYYNYE AGP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

BPNT1, also called bisphosphate 3-prime-nucleotidase, or BPntase, is a member of a magnesium-dependent phosphomonoesterase family. Lithium, a major drug used to treat manic depression, acts as an uncompetitive inhibitor of BPntase. The predicted human protein is 92% identical to mouse BPntase. BPntase's physiologic role in nucleotide metabolism may be regulated by inositol signaling pathways. The inhibition of human BPntase may account for lithium-induced nephrotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPNT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bpnt1
Gene Accession No. O95861
Gene Alias 3'(2'), 5'-bisphosphate nucleotidase 1; 3'(2'),5'-bisphosphate nucleotidase 1; Bisphosphate 3'-nucleotidase 1; BPNT1; BPntase; epididymis luminal protein 20; HEL20; PAP-inositol 1,4-phosphatase; PAP-inositol-1,4-phosphatase; PIP; Sal3; scHAL2 analogous 3; testicular tissue protein Li 29
Gene Symbols Bpnt1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BPNT1. Recombinant protein control fragment (Product #RP-94410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.