Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BPTU
GREENER_CHOICE

Catalog No. p-7114662
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-781-0 10 mg
60-785-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-781-0 Supplier Tocris Bioscience™ Supplier No. 6078/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

P2Y1 allosteric antagonist

BPTU is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 μM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation, nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation.

Specifications

Chemical Name or Material N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N′-[4-(trifluoromethoxy)phenyl]urea
CAS 870544-59-5
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H22F3N3O3
Formula Weight Observed MW: 445.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.