Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Bradford Reagent, Protein Quantification, Liquid, 500 mL
Description
Bradford reagent is an improved formulation for protein quantification with higher tolerance to detergent contamination and reduced protein-to-protein variation.
- Form: Liquid
- Colorimetric detection: Binds to protein, causing a shift in absorption from 465 nm to 595 nm (color change from brown to blue)
- Performance: Excellent linearity across a defined range of protein concentrations
- Storage buffer constituents: 14% Phosphoric acid, 3% Ethanol, 2.3% α-Cyclodextrin hydrate, 0.23% gamma-Cyclodextrin, 0.005% Brilliant Blue G
- Usage: Cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Protein |
| Substrate | Protein |
| Kit Contents | Bradford Reagent (500 mL) |
| Shipping Condition | Ambient |
| Buffer | 14% phosphoric acid, 3% ethanol, 2.3% α-cyclodextrin hydrate, 0.23% gamma-cyclodextrin, 0.005% Brilliant Blue G |
| Detection Location | In-Solution Detection |
| Format | Bottle |
| Form | Liquid |
| Detection Method | Colorimetric |
| Content And Storage | Store at 4°C. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?