Learn More
Description
- Printer Case designed for use with the BMP™41 Label Printer from Brady™
- The durable hard case protects your printer against impacts and dust while allowing for easy mobility
- Quick Charger plugs directly into battery and charges battery from a wall outlet rather than from inside the printer, power supply formatted for North America
- Printer Battery has a long battery life - prints up to 1000 1 in. x 2 in. labels per battery charge
- Enables cordless printer operation so you can print labels where you need them, when you need them
Specifications
Specifications
| Product Type | Rechargeable Battery Pack |
| Color | Gray |
| For Use With (Equipment) | BMP41 Label Printer |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.