Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Accessories for BMP-41 Label Printer

Catalog No. 19000637 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
External Battery Quick Charger
Hard Case
Rechargeable Battery Pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-637 External Battery Quick Charger
19-000-633 Hard Case
19-000-630 Rechargeable Battery Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-637 Supplier Brady™ Supplier No. BMP41QC

Printer case, Quick charger and Printer battery for use with BMP-41 label printer (printer sold separately).

  • Printer Case designed for use with the BMP™41 Label Printer from Brady™
  • The durable hard case protects your printer against impacts and dust while allowing for easy mobility
  • Quick Charger plugs directly into battery and charges battery from a wall outlet rather than from inside the printer, power supply formatted for North America
  • Printer Battery has a long battery life - prints up to 1000 1 in. x 2 in. labels per battery charge
  • Enables cordless printer operation so you can print labels where you need them, when you need them

Specifications

Product Type External Battery Quick Charger
Color Black
For Use With (Equipment) BMP41 Label Printer
Sterility Non-sterile
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.