Learn More
Description
- BMP™51 and BMP™53 Printer Accessories
- Printer Cases designed for use with BMP™51 and BMP™53 Label Printers from Brady™ protects your printer against impacts and dust while allowing for easy mobility
- Printer Magnet Attachment allows printer to affix to panel door or metal surface for more hands-free operation or storage
- Attaches to back of printer with screws to ensure stability
- Printer Battery Tray fits 8 AA batteries
- User may pre-load batteries into the spare tray for quick changeover in the field
- The battery tray snaps in place into the battery compartment
Specifications
Specifications
| Product Type | Soft Case |
| Color | Black |
| For Use With (Equipment) | BMP51 Label Printer, BMP53 Label Printer |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.