Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Accessories for BMP71 Label Printer

Catalog No. 19000207 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Hard Case
Quick Charger
Rechargeable Battery Pack
Soft Case
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-207 Soft Case
19-000-211 Quick Charger
19-000-204 Rechargeable Battery Pack
19-000-206 Hard Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-000-207 Supplier Brady™ Supplier No. M71SC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Printer case, Quick charger and Printer battery for use with BMP-71 Label Printer (printer sold separately).

  • BMP™71 Label Printer Accessories from Brady™
  • Printer Case designed to protect your printer against impacts and dust while allowing for easy mobility.
  • Printer Battery offers long battery life - prints up to 3900 1 in. x 2 in. labels per charge, enables cordless printer operation so you can print labels where you need them, when you need them.
  • BMP™71 Quick Charger, charges battery in one hour.

Specifications

Product Type Soft Case
Color Black
For Use With (Equipment) BMP71 Label Printer
Sterility Non-sterile
Description Soft Carry Case
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.