Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Signs: DANGER ELECTRICAL HAZARD AUTHORIZED PERSONNEL ONLY

Catalog No. 19103662 Shop All Brady Corporation Products
Change view
Click to view available options
Height (Metric):
8.9 cm
17.7 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
19-103-662 8.9 cm
19-835-122 17.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-103-662 Supplier Brady™ Supplier No. 87755
Add to Cart
Add to Cart

For indicating and identifying electrical hazard and high voltage areas

  • Self sticking polyester
      Black/red on white

Specifications

Material Polyester
Quantity 1 Ea.
Description Self-sticking Polyester
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Displayed Text DANGER - ELECTRICAL HAZARD AUTHORIZED PERSONNEL ONLY
Product Type Admittance Sign
Color Black

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.