Learn More
Description
- Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
- Good chemical, oil and water resistance
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Quick application
Specifications
Specifications
| Adhesive Type | Acrylic |
| Color | Yellow |
| Finish | Matte |
| Format | Tape |
| Material | Vinyl |
| Printable Sides | Single-side Printable |
| Product Type | Marking Tape |
| Sterility | Non-sterile |
| For Use With (Application) | For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification |
| Life Span | 3 Years |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.