Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Aisle Marking Tapes

Catalog No. 19016206 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Width (English):
2 in.
3 in.
Quantity:
1 Ea.
1 Roll
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (English) Quantity
19-016206 Green 2 in. 1 Roll
19-046-622 Red 3 in. 1 Roll
19-046-623 Green 3 in. 1 Roll
19-016204 Yellow 2 in. 1 Roll
19-016213 White 3 in. 1 Roll
19-046-621 Black 2 in. 1 Ea.
19-046-625 Black 3 in. 1 Ea.
19-046-624 Blue 3 in. 1 Roll
19-081380 Blue 2 in. 1 Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-016206 Supplier Brady™ Supplier No. 58202
Add to Cart
Add to Cart

Ultra strong B-726 abrasion-resistant vinyl Aisle Marking Tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.

  • Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
  • Good chemical, oil and water resistance
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Quick application

Specifications

Adhesive Type Acrylic
Color Green
Format Tape
Material Vinyl
Product Type Marking Tape
Sterility Non-sterile
For Use With (Application) For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification
Life Span 3 Years
Quantity 1 Roll
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.