Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Barricade Tapes

Catalog No. 19104324 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Orange
Black, Red
Black, Yellow
Length (English):
200 ft.
1000 ft.
Displayed Text:
Blank
Caution
Caution Biohazard
Caution Construction Area
Caution Do Not Enter
Caution High Voltage
Danger
Danger Do Not Enter
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Displayed Text
19-104-324 Black, Yellow 200 ft. Caution Do Not Enter
19-104-322 Black, Yellow 200 ft. Caution Construction Area
19-104-323 Black, Yellow 200 ft. Caution High Voltage
19-104-332 Black, Yellow 1000 ft. Caution
19-104-334 Black, Yellow 1000 ft. Caution Construction Area
19-104-335 Black, Yellow 1000 ft. Blank
19-104-285 Black, Red 1000 ft. Danger Do Not Enter
19-104-318 Black, Red 200 ft. Danger
19-104-319 Black, Red 1000 ft. Danger
19-104-331 Black, Orange 1000 ft. Caution Biohazard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-104-324 Supplier Brady™ Supplier No. 91224
Add to Cart
Add to Cart

For temporary outdoor or permanent indoor use

  • Non-adhesive
  • Lightweight
  • Provides fast, on-the-spot warning
  • Can be tied, stapled or nailed to posts, fences or metal barricades
  • Custom legends also available

Specifications

Dimensions (L x W) 200 ft. x 3 in.
Color Black, Yellow
Material HDPE
Displayed Text Caution Do Not Enter
Length (English) 200 ft.
Length (Metric) 60.9 m
Width (English) 3 in.
Width (Metric) 7.62 cm
Type Barricade Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.