Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard Identification Vinyl Tapes

Catalog No. 19140999 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-140-999 Black, Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-140-999 Supplier Brady™ Supplier No. 88051

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Biohazard Identification Vinyl Tapes are use for identification of biohazards such as bloodborne pathogens.

  • Good resistance to most common chemicals, oils and water
  • Complies with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
  • Visual barricading provides fast on-the-spot warning of potential danger

Specifications

Certifications/Compliance OSHA 1910.144
Adhesive Type Acrylic, Self-adhesive
Color Black, Orange
Material Vinyl (B-933)
Sterility Non-sterile
For Use With (Application) For identifying biohazard areas and bloodborne pathogens
Quantity 1 Ea.
Thickness (English) 0.005 in.
Dimensions (L x W) 32 m x 61 cm (108 ft. x 2 in.)
Length (English) 108 ft.
Length (Metric) 32 m
Width (English) 2 in.
Width (Metric) 61 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.