Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard with Pictogram Signs

Catalog No. 19071419 Shop All Brady Corporation Products
Change view
Click to view available options
Height (English):
3.5 in.
5 in.
7 in.
Width (English):
3.5 in.
5 in.
10 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English)
19-071419 5 in. 3.5 in.
19-807-279 7 in. 10 in.
19-835-103 3.5 in. 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-071419 Supplier Brady™ Supplier No. 89169

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

Bright colors and bold text provide high visibility to critical messages.

 

  • Biohazard marking is required by safety code 1910.1030(g)(1)
  • Designed to withstand harsh industrial environments
  • Black on orange
  • B-485 self-sticking polyester — polyester film with sensitive acrylic

 

Compliance

Meets OSHA Standards 1910.145(e)(4) and 1910.1030(g)(1)

Specifications

Certifications/Compliance OSHA
Material Polyester
Quantity 1 Ea.
For Use With (Application) Used to identify the actual or potential presence of a biologically hazardous material
Height (English) 5 in.
Max. Temperature (English) 122°F
Min. Temperature (English) -40°F
Thickness 0.01 in.
Width (English) 3.5 in.
Temperature Range (Metric) -40°C to 50°C
Temperature Range (English) -40°F to 122°F
Dimensions (L x W) 5 x 3.5 in.
Displayed Text CAUTION Biohazard
Product Type Biohazard Sign
Color Black on Orange
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.