Learn More
Description
Bright colors and bold text provide high visibility to critical messages.
- Biohazard marking is required by safety code 1910.1030(g)(1)
- Designed to withstand harsh industrial environments
- Black on orange
- B-485 self-sticking polyester — polyester film with sensitive acrylic
Compliance
Meets OSHA Standards 1910.145(e)(4) and 1910.1030(g)(1)
Specifications
Specifications
| Certifications/Compliance | OSHA |
| Material | Polyester |
| Quantity | 1 Ea. |
| For Use With (Application) | Used to identify the actual or potential presence of a biologically hazardous material |
| Height (English) | 7 in. |
| Max. Temperature (English) | 122°F |
| Min. Temperature (English) | -40°F |
| Thickness | 0.01 in. |
| Width (English) | 10 in. |
| Temperature Range (Metric) | -40°C to 50°C |
| Show More |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
