Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Blank Hazardous Communication Panels

Catalog No. 19016246 Shop All Brady Corporation Products
Change view
Click to view available options
Width (English):
2.5 in.
5 in.
7.5 in.
10 in.
11 in.
16 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
19-016-246 16 in.
19-016-238 5 in.
19-016-239 7.5 in.
19-016-240 10 in.
19-016-237 2.5 in.
19-016-313 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-016-246 Supplier Brady™ Supplier No. 58510

Requires

Quick-Align™ numbers (0 through 4) and symbols (Oxygen, No Water, Radiation, Acid, Corrosive, and Alkali) needed to customize signs sold separately in sizes from 1 to 6 in. . Contact your Fisher Safety Customer Service Representative for ordering information.

Blank panels to create customized signs

Make signs to identify hazardous chemical containers and storage areas.

  • Place directly on storage tanks, processing tanks and vessels, feed hoppers, pipelines, and other potentially hazardous equipment
  • Upper three corners of signal rate health, fire and reactivity on a scale of 0 to 4; lower corner provides specific hazard information
  • Available in vinyl with permanent pressure-sensitive adhesive or in rigid fiberglass with predrillled holes for mounting to walls

Compliance

Signals meet NFPA Code 704M and Federal Standard 313.

Specifications

Material Fiberglass
Quantity 1 Ea.
Description Premium Fiberglass
Height (English) 16 in.
Width (English) 16 in.
Displayed Text NFPA Diamond
Product Type Hazardous communication label
Color Yellow on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.