Learn More
Brady™ Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
GSA_VA
High adhesion and high pliability are the hallmarks of Brady’s nylon cloth (B-499) label material designed to meet a broad array of identification requirements from control panels to liquid nitrogen environments.
$94.40 - $105.90
Specifications
Autoclavable Autoclavable
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815), UL
Description Aggressive Adhesive Multi-purpose Nylon Labels
Adhesive Type Permanent Acrylic
Color Black, White
View More Specs
Products 3
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19-102-720
GSA_VA
View Documents
Brady™
M21375499
0.3 in.
Each for $94.40
Only null left
Edge
Add to Cart
19-102-733
GSA_VA
View Documents
Brady™
M21500499
0.5 in.
Each for $98.00
Only null left
Add to Cart
19-102-746
GSA_VA
View Documents
Brady™
M21750499
0.75 in.
Each for $105.90
Only null left
Edge
Add to Cart
Description
Description
For wire, cable, faceplates and patch panels
  • High adhesion and high pliability are the hallmarks of Brady’s nylon cloth (B-499) label material designed to meet a broad array of identification requirements from control panels to liquid nitrogen environments.
  • Application Versatility — Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, Polypropylene, and many other material types.
  • Engineered Performance — Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents.
  • Irregular Surfaces — Low profile label material adheres to smooth, rough, or powder-coated surfaces or even materials with low surface energy.
  • Certifications — UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Autoclavable
Aggressive Adhesive Multi-purpose Nylon Labels
Black, White
Cartridge (Label)
Thermal Transfer
Non-sterile
M210;M210-LAB;M211;BMP21-PLUS;BMP21-LAB;IDPAL;LABPAL
-40°C
1 Roll
-40°F to +194°F
6.5 mil
16 ft.
4.8 cm
Halogen-free (DIN VDE 0472/parts 815), UL
Permanent Acrylic
Matte
Nylon Cloth
Label Cartridge
Communications cables, General identification, Laboratory identification, Panel identification, Voice or data identification, Wire and cable marking
1
1 Tape / Box
Rectangle
-40°C to +90°C
0.165 mm
4.88 m
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.