Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradylink™ Striped Warning Posts

Catalog No. 19104363 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
1 Ea.
6/Pk.
Product Type:
Non-reflective Warning Post
Reflective Warning Post
Diameter (English) Base:
14 in.
16 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Product Type Diameter (English) Base
19-104-363 6/Pk. Reflective Warning Post 16 in.
19-104-361 1 Ea. Non-reflective Warning Post 16 in.
19-104-362 6/Pk. Non-reflective Warning Post 16 in.
19-104-356 6/Pk. Non-reflective Warning Post 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-104-363 Supplier Brady™ Supplier No. 92125

Used as decorative alternative to metal posts

  • Lightweight yet tough to withstand years of indoor use in temporary or permanent applications
  • Portable, non-rusting, colorful polystyrene, polyethylene posts and chain
  • Posts are non-conductive so can be safely used in electrically hazardous areas
  • Hollow base can be filled with sand for more stable application
  • Stripes alternate every 10.1cm (4 in.)
  • Large C-hook (sold separately) holds chain in place on large posts
  • Can be used open or closed for greater security
  • Not intended to support heavy weight

Specifications

Color Black, Yellow
Product Type Reflective Warning Post
Height (Metric) 104.1 cm
Material Polyethylene, Polystyrene
Type Reflective Warning Post
Diameter (English) Base 16 in.
Diameter (Metric) Outer 7.6 cm
Height (English) 41 in.
Diameter (English) Outer 3 in.
Diameter (Metric) Base 40.6 cm
Packaging Quantity 6/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.