Learn More
Description
- Package contains replacement filter only
- The manufacturer recommends changing the BSP™31 filter every six months or 5000 cassettes
- Media in filter meets the requirements of a UL Class 1 fire rating
- Dispose of used filters in accordance with local, state and federal regulations (potassium permanganate media)
Specifications
Specifications
| Adhesive Type | Non-adhesive |
| Color | Black |
| Format | Label |
| Printable Sides | Single-side Printable |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Equipment) | Label attachment system |
| Quantity | 1 Ea. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.