Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Chemical and Hazardous Materials Signs: Chlorine

Brady chemical signs are used to identify hazardous material, like Chlorine, to help sustain a safe workplace.

$60.50 - $87.00

Specifications

Grade Industrial
Quantity 1 Ea.
Adhesive Type Non-adhesive
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environment
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-016888
View Documents
Brady™
72408
Fiberglass
Each for $87.00
Only null left
Add to Cart
 
19-807-019
View Documents
Brady™
40878
Aluminum
Each for $60.50
Only null left
Add to Cart
 
Description

Description

  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications

Specifications

Industrial
Non-adhesive
Industrial, utility, commercial and institutional environment
-40°F
35.56 cm
DANGER Chlorine
Red on White
1 Ea.
Surface Mount
10 in.
14 in.
10 x 14 in.
Chemical and Hazardous Materials Sign
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.